
Losartan impurity D

Synonym(s):
2-Butyl-5-chloro-1H-imidazole-4-carboxaldehyde, 2-Butyl-4-chloro-5-formylimidazole
Linear Formula:
C3N2HClCHOCH2CH2CH2CH3
CAS Number:
Molecular Weight:
186.64
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing