
Lorazepam for system suitability

Synonym(s):
(±)-Lorazepam, 7-Chloro-5-(2-chlorophenyl)-1,3-dihydro-3-hydroxy-2H-1,4-benzodiazepin-2-one
Empirical Formula (Hill Notation):
C15H10Cl2N2O2
CAS Number:
Molecular Weight:
321.16
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing