![Levofloxacin Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/388/313/5b1a68a2-b8f2-4586-8cb5-32869f9d1206/640/5b1a68a2-b8f2-4586-8cb5-32869f9d1206.jpg)
Levofloxacin Related Compound C
![Levofloxacin Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/388/313/5b1a68a2-b8f2-4586-8cb5-32869f9d1206/640/5b1a68a2-b8f2-4586-8cb5-32869f9d1206.jpg)
Synonym(s):
(3S)-Ethyl 9-fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylate
Empirical Formula (Hill Notation):
C20H24FN3O4
CAS Number:
Molecular Weight:
389.42
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2