L-Asparagine
Synonym(s):
(S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide
Linear Formula:
H2NCOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
132.12
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Amino Acid
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing