![L-α-Neopentylglycine ≥98.0% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/359/219/6fb2574f-0b50-4e38-8d76-26a276c05ea7/640/6fb2574f-0b50-4e38-8d76-26a276c05ea7.png)
L-α-Neopentylglycine
![L-α-Neopentylglycine ≥98.0% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/359/219/6fb2574f-0b50-4e38-8d76-26a276c05ea7/640/6fb2574f-0b50-4e38-8d76-26a276c05ea7.png)
Synonym(s):
γ-Methyl-L-leucine, (S)-2-Amino-4,4-dimethylpentanoic acid, L-γ-Methylleucine
Empirical Formula (Hill Notation):
C7H15NO2
CAS Number:
Molecular Weight:
145.20
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing