Skip to Content
Merck
Kanamycin Sulfate Pharmaceutical Secondary Standard; Certified Reference Material

Kanamycin sulfate

Kanamycin Sulfate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Kanamycin A sulfate salt, Kanamycin sulfate from Streptomyces kanamyceticus, Kanamycin A, Kanamycin sulfate salt
Empirical Formula (Hill Notation):
C18H36N4O11 · H2O4S
CAS Number:
Molecular Weight:
582.58
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique