Skip to Content
Merck
vibrant-m

Kanamycin monosulfate

vibrant-m
Synonym(s):
Kanamycin A sulfate salt
Empirical Formula (Hill Notation):
C18H36N4O11 · H2O4S
CAS Number:
Molecular Weight:
582.58
EC Number:

Products

Product Category
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing