![Isoxaflutole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/154/360/bfa06632-c087-439f-86b8-c08176d6fcca/640/bfa06632-c087-439f-86b8-c08176d6fcca.png)
Isoxaflutole
![Isoxaflutole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/154/360/bfa06632-c087-439f-86b8-c08176d6fcca/640/bfa06632-c087-439f-86b8-c08176d6fcca.png)
Synonym(s):
5-Cyclopropyl-4-[2-(methylsulfonyl)-4-(trifluoromethyl)benzoyl]isoxazole
Empirical Formula (Hill Notation):
C15H12F3NO4S
CAS Number:
Molecular Weight:
359.32
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2