![Isorhapontigenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/249/987/00bebc2e-c7c2-412c-9026-669a47833999/640/00bebc2e-c7c2-412c-9026-669a47833999.png)
Isorhapontigenin
![Isorhapontigenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/249/987/00bebc2e-c7c2-412c-9026-669a47833999/640/00bebc2e-c7c2-412c-9026-669a47833999.png)
Synonym(s):
(E)- 3′-methoxy-3,4′,5-stilbenetriol, (E)-5-[2-(4-Hydroxy-3-methoxyphenyl)ethenyl]-1,3-benzenediol, 3′-Methoxyresveratrol, Isorhapotigenin, Isorhapotogenin
Empirical Formula (Hill Notation):
C15H14O4
CAS Number:
Molecular Weight:
258.27
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing