
IR-1061

Synonym(s):
4-[2-[2-Chloro-3-[(2,6-diphenyl-4H-thiopyran-4-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-2,6-diphenylthiopyrylium tetrafluoroborate
Empirical Formula (Hill Notation):
C44H34ClS2 · BF4
CAS Number:
Molecular Weight:
749.13
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing