Skip to Content
Merck
IPTG ≥99% (TLC), ≤0.1% Dioxane

IPTG

IPTG ≥99% (TLC), ≤0.1% Dioxane
Synonym(s):
Isopropyl β-D-1-thiogalactopyranoside, IPTG, Isopropyl β-D-thiogalactoside
Empirical Formula (Hill Notation):
C9H18O5S
CAS Number:
Molecular Weight:
238.30
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing