![Iohexol for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/277/262/685e5201-6fbe-4f64-b1c9-3a3166beeda5/640/685e5201-6fbe-4f64-b1c9-3a3166beeda5.png)
Iohexol for peak identification
![Iohexol for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/277/262/685e5201-6fbe-4f64-b1c9-3a3166beeda5/640/685e5201-6fbe-4f64-b1c9-3a3166beeda5.png)
Synonym(s):
5-[N-(2,3-Dihydroxypropyl)acetamido]-2,4,6-triiodo-N,N′-bis(2,3-dihydroxypropyl)isophthalamide
Empirical Formula (Hill Notation):
C19H26I3N3O9
CAS Number:
Molecular Weight:
821.14
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing