![Hydantoin 98%](/deepweb/assets/sigmaaldrich/product/structures/129/852/26f03d58-3040-4a06-86bf-c4b00ce65792/640/26f03d58-3040-4a06-86bf-c4b00ce65792.png)
Hydantoin
![Hydantoin 98%](/deepweb/assets/sigmaaldrich/product/structures/129/852/26f03d58-3040-4a06-86bf-c4b00ce65792/640/26f03d58-3040-4a06-86bf-c4b00ce65792.png)
Synonym(s):
Hydantoin
Empirical Formula (Hill Notation):
C3H4N2O2
CAS Number:
Molecular Weight:
100.08
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing