Skip to Content
Merck
Haloxyfop-P-methyl PESTANAL®, analytical standard

Haloxyfop-P-methyl

Haloxyfop-P-methyl PESTANAL®, analytical standard
Synonym(s):
(R)-2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridyloxy]phenoxy}propanoic acid methyl ester
Empirical Formula (Hill Notation):
C16H13ClF3NO4
CAS Number:
Molecular Weight:
375.73
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing