![Furilazole PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/418/034/53e55710-8f8a-43cb-80d4-5ebcb4533b5d/640/53e55710-8f8a-43cb-80d4-5ebcb4533b5d.png)
Furilazole
![Furilazole PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/418/034/53e55710-8f8a-43cb-80d4-5ebcb4533b5d/640/53e55710-8f8a-43cb-80d4-5ebcb4533b5d.png)
Synonym(s):
(RS)-3-Dichloroacetyl-5-(2-furanyl)-2,2-dimethyl-1,3-oxazolidine
Empirical Formula (Hill Notation):
C11H13Cl2NO3
CAS Number:
Molecular Weight:
278.13
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing