Skip to Content
Merck
Fumonisin B1 from Fusarium moniliforme ≥98% (HPLC)

Fumonisin B1 from Fusarium moniliforme

Fumonisin B1 from Fusarium moniliforme ≥98% (HPLC)
Synonym(s):
Macrofusine
Empirical Formula (Hill Notation):
C34H59NO15
CAS Number:
Molecular Weight:
721.83

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing