Skip to Content
Merck
Fluoxetine hydrochloride VETRANAL®, analytical standard

Fluoxetine hydrochloride

Fluoxetine hydrochloride VETRANAL®, analytical standard
Synonym(s):
(±)-N-Methyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride, LY-110,140 hydrochloride, Prozac®, Fluoxetine hydrochloride
Empirical Formula (Hill Notation):
C17H18F3NO · HCl
CAS Number:
Molecular Weight:
345.79
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique