Skip to Content
Merck
Flavoxate Related Compound A certified reference material, pharmaceutical secondary standard

Flavoxate Related Compound A

Flavoxate Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid
Empirical Formula (Hill Notation):
C17H12O4
CAS Number:
Molecular Weight:
280.27

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade