Flavanone
Synonym(s):
2,3-Dihydroflavone
Empirical Formula (Hill Notation):
C15H12O2
CAS Number:
Molecular Weight:
224.25
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing