![Famciclovir Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/379/925/40d3dda3-73fc-46a6-9c94-7f0ac6ffc673/640/40d3dda3-73fc-46a6-9c94-7f0ac6ffc673.jpg)
Famciclovir Related Compound B
![Famciclovir Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/379/925/40d3dda3-73fc-46a6-9c94-7f0ac6ffc673/640/40d3dda3-73fc-46a6-9c94-7f0ac6ffc673.jpg)
Synonym(s):
4-(2-Amino-9H-purin-9-yl)-2-(hydroxymethyl)butyl acetate
Empirical Formula (Hill Notation):
C12H17N5O3
Molecular Weight:
279.30
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2