Skip to Content
Merck
vibrant-m

Ethinyl estradiol Related Compound B

vibrant-m
Synonym(s):
19-Nor-17α-pregna-1,3,5(10),9(11)-tetraen-20-yne-3,17-diol, 9,11-Dehydroethinylestradiol
Empirical Formula (Hill Notation):
C20H22O2
CAS Number:
Molecular Weight:
294.39

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1