![Estradiol 3-Methyl Ether Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/136/908/3851e20a-b64b-4d2f-8d59-a25afe3f32d4/640/3851e20a-b64b-4d2f-8d59-a25afe3f32d4.jpg)
Estradiol 3-Methyl Ether
![Estradiol 3-Methyl Ether Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/136/908/3851e20a-b64b-4d2f-8d59-a25afe3f32d4/640/3851e20a-b64b-4d2f-8d59-a25afe3f32d4.jpg)
Synonym(s):
β-Estradiol 3-methyl ether, 1,3,5(10)-Estratriene-3,17β-diol 3-methyl ether, 17β-Hydroxy-3-methoxy-1,3,5(10)-estratriene
Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing