Ergometrine maleate
Synonym(s):
Ergonovine maleate salt, Ergometrine maleate salt
Empirical Formula (Hill Notation):
C19H23N3O2 · C4H4O4
CAS Number:
Molecular Weight:
441.48
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing