Skip to Content
FR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(−)-Epicatechin
Synonym(s):
(−)-
cis
-3,3′,4′,5,7-Pentahydroxyflavane, (2
R
,3
R
)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2
H
)-benzopyran-3,5,7-triol, (−)-Epicatechin
Empirical Formula (Hill Notation):
C
15
H
14
O
6
CAS Number:
490-46-0
Molecular Weight:
290.27
EC Number:
207-710-1
Products
Shipping
Ships Today (1)
Product Category
antioxidants (2)
bioactive small molecules (2)
flavonoids (2)
natural products (2)
polyphenols (2)
metabolites (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (2)
primary reference standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
E1753
(−)-Epicatechin,
≥90% (HPLC)
Expand
View Pricing
E4018
(−)-Epicatechin,
≥98% (HPLC), from green tea
Expand
View Pricing
PHL89192
(−)-Epicatechin,
phyproof
®
Reference Substance
Expand
View Pricing
68097
(−)-Epicatechin,
analytical standard
Expand
View Pricing
03940590
(−)-Epicatechin,
primary reference standard
Expand
View Pricing