
Echinocystic acid

Synonym(s):
3β,16α-dihydroxyolean-12-en-28-oic acid
Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing