Dosulepin hydrochloride
Synonym(s):
Dothiepin hydrochloride, 3-Dibenzo[b,e]thepin-11(6H)-ylidine-N,N-dimethyl-1-propanamine hydrochloride
Empirical Formula (Hill Notation):
C19H21NS·HCl
CAS Number:
Molecular Weight:
331.90
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing