![(±)-Lisofylline analytical standard](/deepweb/assets/sigmaaldrich/product/structures/288/088/df301c83-da24-42c1-aa36-13773fe07520/640/df301c83-da24-42c1-aa36-13773fe07520.png)
(±)-Lisofylline
![(±)-Lisofylline analytical standard](/deepweb/assets/sigmaaldrich/product/structures/288/088/df301c83-da24-42c1-aa36-13773fe07520/640/df301c83-da24-42c1-aa36-13773fe07520.png)
Synonym(s):
Lisofylline, (±)-1-(5-Hydroxyhexyl)-3,7-dimethylxanthine, 5-Hydroxy pentoxifylline, Penthydroxyfylline
Empirical Formula (Hill Notation):
C13H20N4O3
CAS Number:
Molecular Weight:
280.32
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing