Skip to Content
Merck
(±)-α-Tocopherol tested according to Ph. Eur.

(±)-α-Tocopherol

(±)-α-Tocopherol tested according to Ph. Eur.
Synonym(s):
DL-all-rac-α-Tocopherol, Vitamin E
Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3