Skip to Content
Merck
(±)-8-Hydroxy-2-(dipropylamino)tetralin hydrobromide ≥98%

(±)-8-Hydroxy-2-(dipropylamino)tetralin hydrobromide

(±)-8-Hydroxy-2-(dipropylamino)tetralin hydrobromide ≥98%
Synonym(s):
(±)-2-Dipropylamino-8-hydroxy-1,2,3,4-tetrahydronaphthalene, (±)-8-Hydroxy-2-(dipropylamino)tetralin, (±)-8-OH-DPAT
Empirical Formula (Hill Notation):
C16H25NO · HBr
CAS Number:
Molecular Weight:
328.29

Products

Shipping
Product Category
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing