Skip to Content
Merck
di-Sodium oxalate for analysis EMSURE®

di-Sodium oxalate

di-Sodium oxalate for analysis EMSURE®
Synonym(s):
di-Sodium oxalate
Empirical Formula (Hill Notation):
C2Na2O4
CAS Number:
Molecular Weight:
134.00

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Technique