
Dimethyl-d6 phthalate

Empirical Formula (Hill Notation):
C10D6H4O4
CAS Number:
Molecular Weight:
200.22
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing