Skip to Content
Merck
Dilithium phthalocyanine Dye content 70 %

Dilithium phthalocyanine

Dilithium phthalocyanine Dye content 70 %
Synonym(s):
29H,31H-Phthalocyanine dilithium salt
Empirical Formula (Hill Notation):
C32H16Li2N8
CAS Number:
Molecular Weight:
526.41

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing