Dibenzo-24-crown-8
Synonym(s):
2,3,14,15-Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosa-2,14-diene, 6,7,9,10,12,13,20,21,23,24,26,27-Dodecahydrodibenz[b,n]-1,4,7,10,13,16,19,22-octaoxacyclotetracosane
Empirical Formula (Hill Notation):
C24H32O8
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing