Deoxyribonucleic acid from calf thymus
Synonym(s):
Calf Thymus DNA, DNA from calf thymus, Thymonucleic acid
CAS Number:
EC Number:
Products
Product Category
Brand
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing