Skip to Content
Merck
Deferasirox CRS, European Pharmacopoeia (EP) Reference Standard

Deferasirox

Deferasirox CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4-[3,5-bis(2-Hydroxyphenyl)-1H-1,2,4-triazol-1-yl]benzoic acid
Empirical Formula (Hill Notation):
C21H15N3O4
CAS Number:
Molecular Weight:
373.36

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Research Area
API Family
Showing 1-2 of 2
Compare
Product Number
Description
Pricing