Skip to Content
Merck
(+)-1,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene kanata purity

(+)-1,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene

(+)-1,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene kanata purity
Synonym(s):
(2S,2′S,5S,5′S)-2,2′,5,5′-Tetramethyl-1,1′-(o-phenylene)diphospholane, (S,S)-Me-DUPHOS, (S,S)-Methyl-DUPHOS
Empirical Formula (Hill Notation):
C18H28P2
CAS Number:
Molecular Weight:
306.36

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing