Skip to Content
Merck
Copper(II) acetylacetonate ≥99.9% trace metals basis

Copper(II) acetylacetonate

Copper(II) acetylacetonate ≥99.9% trace metals basis
Synonym(s):
2,4-Pentanedione copper(II) derivative, Bis(2,4-pentanedionato)copper(II), Cu(acac)2, Cupric acetylacetonate, Copper(II) acetylacetonate
Linear Formula:
Cu(C5H7O2)2
CAS Number:
Molecular Weight:
261.76

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-3 of 3
Compare
Product Number
Description
Pricing