Skip to Content
Merck
vibrant-m

Clomipramine impurity C

vibrant-m
Synonym(s):
3-Chloro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine, 3-(3-Chloro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine
Empirical Formula (Hill Notation):
C19H21ClN2
CAS Number:
Molecular Weight:
312.84

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing