Skip to Content
Merck
Clidinium Bromide Related Compound A certified reference material, pharmaceutical secondary standard

Clidinium bromide Related Compound A

Clidinium Bromide Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
3-Hydroxy-1-methylquinuclindinium bromide
Empirical Formula (Hill Notation):
C8H16BrNO
Molecular Weight:
222.12

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade