![Cisatracurium besylate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/408/444/ffaf1fc0-9fd2-41a2-b50b-1f99289a03c4/640/ffaf1fc0-9fd2-41a2-b50b-1f99289a03c4.png)
Cisatracurium besylate
![Cisatracurium besylate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/408/444/ffaf1fc0-9fd2-41a2-b50b-1f99289a03c4/640/ffaf1fc0-9fd2-41a2-b50b-1f99289a03c4.png)
Empirical Formula (Hill Notation):
C53H72N2O12·2C6H5O3S
CAS Number:
Molecular Weight:
1243.48
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing