Skip to Content
Merck
cis-11,14-Eicosadienoic acid ≥98%, liquid

cis-11,14-Eicosadienoic acid

cis-11,14-Eicosadienoic acid ≥98%, liquid
Linear Formula:
CH3(CH2)4(CH=CHCH2)2(CH2)8CO2H
CAS Number:
Molecular Weight:
308.50

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing