Cidofovir hydrate
Synonym(s):
(S)-1-[3-hydroxy-2-(phosphonylmethoxy)propyl]cytosine, (S)-HPMPC; Cidovir, HPMPC, Vistide
Empirical Formula (Hill Notation):
C8H14N3O6P · xH2O
CAS Number:
Molecular Weight:
279.19 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing