Cholesterol-3,4-13C2
Empirical Formula (Hill Notation):
13C2C25H46O
CAS Number:
Molecular Weight:
388.64
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Special Grade
Technique
Showing 1-3 of 3