Skip to Content
Merck
Cetirizine dihydrochloride European Pharmacopoeia (EP) Reference Standard

Cetirizine dihydrochloride

Cetirizine dihydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
[2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetic acid dihydrochloride
Empirical Formula (Hill Notation):
C21H25ClN2O3 · 2HCl
CAS Number:
Molecular Weight:
461.81

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Relevant Disease
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing