![Cerium(III) bromide AnhydroBeads™, −10 mesh, 99.99% trace metals basis](/deepweb/assets/sigmaaldrich/product/structures/338/927/a3003b2c-6575-4020-901f-e4c60d65bbe3/640/a3003b2c-6575-4020-901f-e4c60d65bbe3.png)
Cerium(III) bromide
![Cerium(III) bromide AnhydroBeads™, −10 mesh, 99.99% trace metals basis](/deepweb/assets/sigmaaldrich/product/structures/338/927/a3003b2c-6575-4020-901f-e4c60d65bbe3/640/a3003b2c-6575-4020-901f-e4c60d65bbe3.png)
Synonym(s):
Cerium tribromide, Cerous bromide
Linear Formula:
CeBr3
Products
Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing