Skip to Content
Merck
vibrant-m

Cefpodoxime proxetil for impurity H identification

vibrant-m
Synonym(s):
Cefpodoxime proxetil
Empirical Formula (Hill Notation):
C21H27N5O9S2
CAS Number:
Molecular Weight:
557.60

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1