Cefepime Related Compound D
Synonym(s):
(Z)-2-Amino-α-(methoxyimino)-4-thiazoleacetic acid
Empirical Formula (Hill Notation):
C6H7N3O3S
CAS Number:
Molecular Weight:
201.20
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2