Skip to Content
Merck
Cecropin A ≥97% (HPLC), powder

Cecropin A

Cecropin A ≥97% (HPLC), powder
Empirical Formula (Hill Notation):
C184H313N53O46
CAS Number:
Molecular Weight:
4003.78

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing