![vibrant-m](/assets/images/pdp-no-image-sq/pdp-no-image-sq_w640.png)
Captopril disulfide
![vibrant-m](/assets/images/pdp-no-image-sq/pdp-no-image-sq_w640.png)
Synonym(s):
1,1′-[Dithiobis[(2S)-2-methyl-1-oxo-3,1-propanediyl]]bis-L-proline
Empirical Formula (Hill Notation):
C18H28N2O6S2
CAS Number:
Molecular Weight:
432.55
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing