Cannabinoid Mixture (Neutrals) - 8 Component solution
Empirical Formula (Hill Notation):
C19H26O2
Molecular Weight:
286.41
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Chemical Composition
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing